Gas washing bottles, with extra coarse fritted disc, PYREX®
Supplier: Corning
31760-125C 31760-125EC 31760-250C 31760-250EC 31760-500BO 31760-500C 31760-500EC
CORN31760-125CEA 370 SGD
CORN31760-125C CORN31760-125EC CORN31760-250C CORN31760-250EC CORN31760-500BO CORN31760-500C CORN31760-500EC
Gas washing bottles, with extra coarse fritted disc, PYREX®
Bottles Gas Washing Bottles
PYREX®, borosilicate glass.
  • Large hexagonal base
  • Humidify carbon dioxide-air mixtures for maintaining high humidity
  • Fritted disc provides greater efficiency and more uniform dispersion of gas bubbles for complete absorption
These gas washing bottles are used for washing and drying gases. NS 40/50 stopper is used to seal the top to the base. The NS stopper incorporates a tube with a 20 mm diameter extra coarse fritted disc as the distributor. Tubulation outer dimension is 8 mm. This extra coarse frit has a nominal pore distribution of 170 to 220 µm.