VWR® Thermotech, Beakers, PTFE
Supplier: VWR Collection
89026-010 89026-016 89026-024
VWRU89026-010EA 419 SGD
VWRU89026-010 VWRU89026-016 VWRU89026-024
VWR® Thermotech, Beakers, PTFE
Beakers
PTFE, with spout.
  • Improved heat transfer
  • Heatable to 270 °C without distortion
  • Totally inert pure PTFE inner surface
  • Isostatically molded in pure, FDA approved
Chemically resistant and inert beaker which features the unique combination of a pure PTFE body with a specially formulated stabilised PTFE Carbon base as a single molding.