Stoppers for [ST] Flask PYREX®
Supplier: Corning
7650-13 7650-16 7650-19 7650-22 7650-27 7650-32 7650-38 7650-8 7650-9
CORN7650-13EA 75 SGD
CORN7650-13 CORN7650-16 CORN7650-19 CORN7650-22 CORN7650-27 CORN7650-32 CORN7650-38 CORN7650-8 CORN7650-9
Stoppers for [ST] Flask PYREX®
Stoppers Glass Stoppers
These PYREX® hollow borosilicate glass stoppers are light weight, strong to reduce chipping and breakage. Barrel shaped head makes it easy to clean and rotate in or out. Grooves in the sides help prevent slippage.
  • Interchangeable with standard taper joints
Order Now
SPECIFICATIONS
Environmentally Preferable