VWR®, Tilt measures, automatic
Supplier: VWR Collection
53481-213 53481-155 53481-122
VWRU53481-213EA 853 SGD
VWRU53481-213 VWRU53481-155 VWRU53481-122
VWR®, Tilt measures, automatic
Bottle Top Tilt Measures
Dispensers made from Borosilicate glass 3.3 are recommended for multiple test procedures as well as dispensing strong acids, alkalies, and volatile reagents.
  • Accurate to ±2%
  • The calibrated pipette heads automatically fill and deliver contents by simple pouring action for rapid repeat transfers of fixed volumes of solutions
  • Tilting dispenser forward delivers the calibrated volume
Delivery information: A complete unit consists of an interchangeable glass pipette head, Erlenmeyer flask with ground joint, and poly joint clamp. Pipette heads with innerMore Product Information