VWR®, Mortars without pestles, porcelain
Supplier: VWR Collection
89038-144 89038-154 89038-148 89038-150 89038-152 89038-146
VWRU89038-144EA 30.8 SGD
VWRU89038-144 VWRU89038-154 VWRU89038-148 VWRU89038-150 VWRU89038-152 VWRU89038-146
VWR®, Mortars without pestles, porcelain
Mortars and Pestles Mortars
Porcelain, glazed exterior, with spout.
Ordering information: Pestle must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable