Chemistry kit, PYREX®
Supplier: Corning
8950-24 9021-24 9601-24 2401-24 2490-24 6949 6949N-BO 6949N-IO
CORN8950-24EA 181 SGD
CORN8950-24 CORN9021-24 CORN9601-24 CORN2401-24 CORN2490-24 CORN6949 CORN6949N-BO CORN6949N-IO
Chemistry kit, PYREX®
Microchemistry Kits
Chemistry kit is designed for most common organic chemistry experiments.
It comprises 15 components with 24/40 standard tapered joints including flasks, adapters, condensers, condenser column, funnel, stopper and reaction vessel. The components are held in foam within a polyethylene case.
Order Now
SPECIFICATIONS
Environmentally Preferable