Chemistry kit, PYREX®
Supplier: Corning
7815-19 9050-19 6949E 6949E-BO 6949E-IO
CORN7815-19EA 6.6 SGD
CORN7815-19 CORN9050-19 CORN6949E CORN6949E-BO CORN6949E-IO
Chemistry kit, PYREX®
Microchemistry Kits
The chemistry kit is designed for most common organic chemistry experiments.
It comprises 17 components with 19/22 standard tapered joints including distilling column, condenser, round bottom boiling flasks, separatory funnel, stopper, thermometer holder, bleed tube, tube adapter and connecting tube. The components are held in foam within a polyethylene case.