VWR®, Weighing Boats
Supplier: VWR Collection
30287-040 30287-029
VWRU30287-040EA 8350 SGD
VWRU30287-040 VWRU30287-029
VWR®, Weighing Boats
Boats Weighing Boats
Thin-walled glass.
  • Very good chemical resistance
  • Good temperature resistance
  • Flattened base provides stable rest on balance pan
Ideal for speedy, precise weighing and transfer of samples without loss.