Safety cabinets for flammable wastes, Sure-Grip®
Supplier: JUSTRITE MANUFACTURING
8930053 8930253
JUSU8930053EA 2120 SGD
JUSU8930053 JUSU8930253
Safety cabinets for flammable wastes, Sure-Grip®
Furniture Storage Cabinets
Safety cabinets are used for temporary on-site storage solution for flammable waste.
  • Durable, powder-coat finish and chemical resistance
  • Large 'Flammable Waste' warning label on door identifies contents
  • Welded, 1 mm double-walled steel with 38 mm insulating air space for fire resistance
Cabinets feature adjustable SpillSlope® shelves that direct any spills to back and bottom of a leakproof 51 mm sump. Three-point stainless steel bullet latch provides fail-safe, positive door closure.
It also contains dual 51 mm vents with integral flame arresters,More Product Information