VWR®, Storage Bags
Supplier: VWR Collection
89069-100 89069-678
VWRU89069-100EA 26.6 SGD
VWRU89069-100 VWRU89069-678
VWR®, Storage Bags
Bags Storage Bags
These flat bags, in clear LDPE are made with an extra strong bottom seal or a durable side weld which acts as an outstanding vapour and moisture barrier. Ideal for product presentation.
Certifications: Meet FDA and USDA specifications.