Safety storage cabinets for hazardous chemicals
Supplier: JUSTRITE MANUFACTURING
24120 24140
JUSU24120EA 1990 SGD
JUSU24120 JUSU24140
Safety storage cabinets for hazardous chemicals
Furniture Storage Cabinets
Long lasting wood construction along with stainless steel handles and hinges to store highly corrosive liquids such as phenol, nitric acid and sulfuric acid in these wood laminate storage cabinets.
  • Ideal for smaller quantities used at workbench level
  • Shelves adjust on 32 mm centers to accommodate a wide variety of containers
  • Cabinets include adjustable leg levelers, patent pending Haz-Alert™ reflective labels, and double key set for safety and security
Research environments using many different types of chemicals have special storage requirements. Whether using acids, bases or solvents, it is critical that incompatible liquids are segregated to avoid disastrous explosions or fire.