VWR®, Dry block/incubator thermometers
Supplier: VWR Collection
89095-806 89095-802
VWRU89095-806EA 63.1 SGD
VWRU89095-806 VWRU89095-802
VWR®, Dry block/incubator thermometers
Thermometers Analogue Thermometers
These thermometers are used in all standard heating blocks and water baths. It features a PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage.
Red organic liquid filling, against white back glass.