Hybridisation tubes, PYREX®
Supplier: Corning
7995-150 7995-300
CORN7995-150EA 142 SGD
CORN7995-150 CORN7995-300
Hybridisation tubes, PYREX®
Bottles Hybridisation Bottles/Tubes
PYREX® borosilicate glass, GL 45 orange plug-seal cap with silicone O-ring.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These tubes with three different lenghths are used in hybridisation incubators with rotator.
An optional Corning® vented membrane cap is available for low temperature assays.