Polyethylene glycol 200, liquid, J.T.Baker®

Supplier: Avantor

Synonyms: PEG 200, Ethylene glycol polymer average Mn 200

BAKRU214-07EA 102 SGD
BAKRU214-07 BAKRU214-09
Polyethylene glycol 200, liquid, J.T.Baker®
Polyethylene glycol 200
Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: –55…–40 °C
Density: 1,124 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD01779596
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR