Vertical electrophoresis system, omniPAGE WAVE Maxi
Supplier: CLEAVER SCIENTIFIC
VS20WAVECES VS20WAVECES-HI
CLEVVS20WAVECESEA 11970 SGD
CLEVVS20WAVECES CLEVVS20WAVECES-HI
Vertical electrophoresis system, omniPAGE WAVE Maxi
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The omniPAGE WAVE complete electrophoresis system provides a fully integrated solution using one universal gel tank and lid for vertical PAGE, tube gel IEF and electroblotting. Interchangeable modular insert users to combine vertical PAGE with capillary gel IEF to perform 2-D electrophoresis, followed by western transfer for enhanced sensitivity.
  • Cooling supplied for temperature-sensitive separations and samples
  • High-intensity blotting system available for rapid transfers
Delivery information: Each complete system includes WAVE maxi vertical unit incl.More Product Information