Dichloro(ethylenediamine)platinum(II) (≥59,7% Pt)

Supplier: Thermo Scientific
Warning

10497.01 10497.03
ALFA10497.01EA 0 SGD
ALFA10497.01 ALFA10497.03
Dichloro(ethylenediamine)platinum(II) (≥59,7% Pt)
Dichloro(ethylenediamine)platinum(II)

CAS No.: 14096-51-6 Molecular Formula: PtCl2(H2NCH2CH2NH2) Formula Weight: 326.10 Physical Form: Crystalline/Powder Sensitivity: Hygroscopic MDL No.: MFCD00000011

Formula: C₂H₈Cl₂N₂Pt
MW: 326,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000011
CAS Number: 14096-51-6
EINECS: 237-943-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR