Calcium molybdate ≥99.9965% (metals basis), Puratronic®

Supplier: Thermo Scientific
Warning

10682.06 10682.14
ALFA10682.06EA 0 SGD
ALFA10682.06 ALFA10682.14
Calcium molybdate ≥99.9965% (metals basis), Puratronic®
Calcium molybdate

Purity: 99.9965% CAS No.: 7789-82-4 Molecular Formula: CaMoO4 Formula Weight: 200.01 Physical Form: Powder Density: 4.38-4.53 Odour: Odourless Melting Point: 965° MDL No.: MFCD00015976

Formula: CaMoO₄
MW: 200,024 g/mol
Melting Pt: 965 °C
Density: 4,38…4.53
Storage Temperature: Ambient
MDL Number: MFCD00015976
CAS Number: 7789-82-4
EINECS: 232-192-9
Merck Index: 14,1681

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR