Platinum Rhodium Palladium ≥99.9% (metals basis), gauze 80 mesh knitted from 0.076mm (0.003in) dia wire
Supplier: Thermo Scientific
12216.FI 12216.GH
ALFA12216.FIEA 0 SGD
ALFA12216.FI ALFA12216.GH
Platinum Rhodium Palladium ≥99.9% (metals basis), gauze 80 mesh knitted from 0.076mm (0.003in) dia wire
Platinum Rhodium Palladium
Purity: 99.9% Molecular Formula: Pt:Rh:Pd; 90:5:5 wt% Physical Form: ≈0.39g/25x25mm MDL No.: MFCD00011179