Rhodium(III) nitrate solution (10 - 15% Rh) (w/w)

Supplier: Thermo Scientific
Danger

Synonyms: Rhodium(III) nitrate

12633.NA 12633.NJ
ALFA12633.NAEA 0 SGD
ALFA12633.NA ALFA12633.NJ
Rhodium(III) nitrate solution (10 - 15% Rh) (w/w)
Rhodium (III) nitrate

CAS No.: 10139-58-9 Molecular Formula: N3O9Rh Formula Weight: 288.92 Physical Form: Liquid Appearance: Red-brown Odour: Acidic UN No.: UN3093 MDL No.: MFCD00011202

Formula: N₃O₉Rh
MW: 288,92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011202
CAS Number: 10139-58-9
EINECS: 233-397-6
UN: 3093
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR