Calcium metaborate ≥99.992% (metals basis)

Supplier: Thermo Scientific
Warning

12988.06 12988.14 12988.22
ALFA12988.06EA 0 SGD
ALFA12988.06 ALFA12988.14 ALFA12988.22
Calcium metaborate ≥99.992% (metals basis)
Calcium metaborate

Purity: 99.992% CAS No.: 13701-64-9 Molecular Formula: B2CaO4 Formula Weight: 125.70 Physical Form: Powder Appearance: White Odour: Odourless MDL No.: MFCD00049397

Formula: B₂CaO₄
MW: 125,70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049397
CAS Number: 13701-64-9
EINECS: 237-224-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR