Thulium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific

Synonyms: Thulium(III) acetylacetonate

13164.03 13164.06
ALFA13164.03EA 0 SGD
ALFA13164.03 ALFA13164.06
Thulium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Thulium(III) 2,4-pentanedionate

Purity: 99.9% CAS No.: 14589-44-7 Molecular Formula: C15H21O6Tm Formula Weight: 466.26 Physical Form: Powder Odour: Odourless MDL No.: MFCD00058840

Formula: C₁₅H₂₁O₆Tm
MW: 466,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058840
CAS Number: 14589-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR