Lanthanum(III) oxalate decahydrate ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Warning
|
13196.18
13196.30
ALFA13196.18EA
0
SGD
ALFA13196.18
ALFA13196.30
Lanthanum(III) oxalate decahydrate ≥99.99% (REO, rare earth oxide basis)
Lanthanum(III) oxalate decahydrate
Purity: 99.99%
CAS No.: 537-03-1
Molecular Formula: C6La2O12.10H2O
Formula Weight: 722.02 (541.87anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: dec.
UN No.: UN3288
MDL No.: MFCD00150540
Formula:
C₆La₂O₁₂·10H₂O MW: 722,02 g/mol (541,87 anhy) Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00150540 CAS Number: 537-03-1 EINECS: 208-656-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...