Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 304
Supplier: Thermo Scientific
13476.CH
13476.RR
13476.RU
ALFA13476.CHEA
0
SGD
ALFA13476.CH
ALFA13476.RR
ALFA13476.RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...