1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
13932.04
13932.09
ALFA13932.04EA
92.2
SGD
ALFA13932.04
ALFA13932.09
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥98%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
Purity: 98%
CAS No.: 14647-23-5
Molecular Formula: NiCl2[(C6H5)2PCH2CH2P(C6H5)2]
Formula Weight: 528.02
Physical Form: Orange-red to brown powder
Appearance: Orange-red to brown
Melting Point: 263-265°
MDL No.: MFCD00013313
Formula:
C₂₆H₂₄Cl₂NiP₂ MW: 528,016 g/mol Melting Pt: 263…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013313 CAS Number: 14647-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...