Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Warning
|
ampouled under argon
38585.03
38585.06
ALFA38585.03EA
0
SGD
ALFA38585.03
ALFA38585.06
Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (REO, rare earth oxide basis)
Tris(cyclopentadienyl)gadolinium(III)
Purity: 99.9%
CAS No.: 1272-21-5
Molecular Formula: C15H15Gd
Formula Weight: 352.54
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
UN No.: UN3181
MDL No.: MFCD00145476
Formula:
C₁₅H₁₅Gd Storage Temperature: Ambient |
MDL Number:
MFCD00145476 CAS Number: 1272-21-5 EINECS: 215-046-9 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...