Cyclopentadienylniobium(V) tetrachloride
Supplier: Thermo Scientific
|
Danger
|
39246.03
39246.MD
ALFA39246.03EA
0
SGD
ALFA39246.03
ALFA39246.MD
Cyclopentadienylniobium(V) tetrachloride
Cyclopentadienylniobium(V) tetrachloride
CAS No.: 33114-15-7
Molecular Formula: C5H5Cl4Nb
Formula Weight: 299.82
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Melting Point: 180° dec.
UN No.: UN3261
MDL No.: MFCD00070445
Formula:
C₅H₅Cl₄Nb MW: 299,801 g/mol Melting Pt: 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070445 CAS Number: 33114-15-7 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...