Iron(III) 1,1,1-trifluoro-2,4-pentanedionate

Supplier: Thermo Scientific
Warning

Synonyms: Trifluoroacetylacetono Iron(III) salt, Ferric(III) trifluoroacetylacetonate, Tris(trifluoro-2,4-pentanedionato)iron(III), Iron(III) trifluoroacetylacetonate

39265.06 39265.14
ALFA39265.06EA 0 SGD
ALFA39265.06 ALFA39265.14
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate

CAS No.: 14526-22-8 Molecular Formula: C15H12F9FeO6 Formula Weight: 515.09 Physical Form: Crystalline Melting Point: 110-112° MDL No.: MFCD00017760

Formula: C₁₅H₁₂F₉FeO₆
MW: 515,086 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00017760
CAS Number: 14526-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR