Tris(cyclopentadienyl)neodymium(III) ≥99.5%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Tris(cyclopentadienyl)neodymium
ampouled under argon
39300.03
39300.06
ALFA39300.03EA
0
SGD
ALFA39300.03
ALFA39300.06
Tris(cyclopentadienyl)neodymium(III) ≥99.5%
Tris(cyclopentadienyl)neodymium(III)
Purity: 99.5%
CAS No.: 1273-98-9
Molecular Formula: C15H15Nd
Formula Weight: 339.53
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 417°
Boiling Point: 220°/0.01mm subl.
UN No.: UN3181
MDL No.: MFCD00145477
Formula:
C₁₅H₁₅Nd MW: 339,522 g/mol Boiling Pt: 220 °C (0,01 mmHg) (subl.) Melting Pt: 417 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145477 CAS Number: 1273-98-9 EINECS: 215-057-9 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...