Iron yttrium oxide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific
Warning

39488.14 39488.22
ALFA39488.14EA 0 SGD
ALFA39488.14 ALFA39488.22
Iron yttrium oxide ≥99.9% (REO, rare earth oxide basis)
Iron yttrium oxide

Purity: 99.9% CAS No.: 12063-56-8 Molecular Formula: Y3Fe5O12 Formula Weight: 737.94 Physical Form: Powder Odour: Odourless MDL No.: MFCD00210618

Formula: Y₃Fe₅O₁₂
MW: 737,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210618
CAS Number: 12063-56-8
EINECS: 235-053-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR