Barium calcium copper oxide (2-2-3) ≥99.9% (metals basis), fine agglomerate
Supplier: Thermo Scientific
|
Warning
|
40122.09
40122.18
ALFA40122.09EA
0
SGD
ALFA40122.09
ALFA40122.18
Barium calcium copper oxide (2-2-3) ≥99.9% (metals basis), fine agglomerate
Barium calcium copper oxide
Purity: 99.9%
Molecular Formula: Ba2Ca2Cu3Ox, Multiple Phases
Physical Form: Fine Agglomerate
UN No.: UN1564
MDL No.: MFCD00211652
Formula:
Ba₂CaCu₂Ox,MultiplePhases Storage Temperature: Ambient |
MDL Number:
MFCD00211652 UN: 1564 ADR: 6.1,III |
Learn more
![](/m-sg.vwr.com/en_SG/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...