Silver hydrogen fluoride ≥99%

Supplier: Thermo Scientific
Danger

40129.09 40129.18
ALFA40129.09EA 0 SGD
ALFA40129.09 ALFA40129.18
Silver hydrogen fluoride ≥99%
Silver hydrogen fluoride

Purity: 99% min CAS No.: 12249-52-4 Molecular Formula: AgF2H Formula Weight: 146.87 Physical Form: Crystalline Sensitivity: Light Sensitive; Hygroscopic Melting Point: dec. UN No.: UN1740 MDL No.: MFCD00042145

Formula: AgF₂H
MW: 146,87 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00042145
CAS Number: 12249-52-4
UN: 1740
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR