Polyethyleneimine, M.W. 10,000 ≥99%, branched
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
40331.14
40331.22
40331.36
ALFA40331.14EA
103
SGD
ALFA40331.14
ALFA40331.22
ALFA40331.36
Polyethyleneimine, M.W. 10,000 ≥99%, branched
Polyethyleneimine
Purity: 99%
CAS No.: 9002-98-6
Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y
Physical Form: Liquid
Appearance: Pale yellow
Odour: Amine-like
MDL No.: MFCD00803910
Formula:
(CH₂CH₂NH)n Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...