Sodium tetracyanoplatinate(II) hydrate (≥48,4% Pt) ≥99.95% (metals basis), Premion®
Supplier: Thermo Scientific
|
Warning
|
40400.03
40400.06
ALFA40400.03EA
0
SGD
ALFA40400.03
ALFA40400.06
Sodium tetracyanoplatinate(II) hydrate (≥48,4% Pt) ≥99.95% (metals basis), Premion®
Sodium tetracyanoplatinate(II) hydrate
Purity: 99.95%
CAS No.: 699012-94-7
Molecular Formula: Na2Pt(CN)4.xH2O (x≈3)
Formula Weight: 345.14(anhy)
Physical Form: Crystalline powder
Appearance: White
Melting Point: Dec.
MDL No.: MFCD00798527
Formula:
Na₂Pt(CN)₄·xH₂O(x≈3) MW: 345,14 g/mol (anhy) Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00798527 CAS Number: 699012-94-7 EINECS: 239-536-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...