Bismuth (III) tert-pentyloxide ≥97%
Supplier: Thermo Scientific
|
Warning
|
41203.03
41203.06
ALFA41203.03EA
0
SGD
ALFA41203.03
ALFA41203.06
Bismuth (III) tert-pentyloxide ≥97%
Bismuth (III) tert-pentyloxide
Purity: 97+%
CAS No.: 124687-44-1
Molecular Formula: C15H33BiO3
Formula Weight: 470.41
Physical Form: Powder
Appearance: Dark grey
Melting Point: 180° dec.
Boiling Point: 63°/10-4mm subl.
UN No.: UN3181
MDL No.: MFCD00151666
Formula:
C₁₅H₃₃BiO₃ MW: 470,41 g/mol Boiling Pt: 63 °C (10⁴ mmHg) (subl.) Melting Pt: 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00151666 CAS Number: 124687-44-1 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...