Aluminum lanthanum isopropoxide 7% (w/v) in isopropanol (IPA) , 99% (metals basis)

Supplier: Thermo Scientific
Danger

41262.AC 41262.AE
ALFA41262.ACEA 0 SGD
ALFA41262.AC ALFA41262.AE
Aluminum lanthanum isopropoxide 7% (w/v) in isopropanol (IPA) , 99% (metals basis)
Aluminum lanthanum isopropoxide

Purity: 99% CAS No.: 33939-97-8 Molecular Formula: La[Al(OCH(CH3)2)4]3 Formula Weight: 928.24 Physical Form: Liquid Sensitivity: Moisture Sensitive Boiling Point: 208°/0.5mm UN No.: UN1219 MDL No.: MFCD00143591


Applications: Thin films of metal oxides

Formula: La[Al(OCH(CH₃)₂)₄]₃
MW: 928,24 g/mol
Boiling Pt: 208 °C (0,5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00143591
CAS Number: 33939-97-8
UN: 1219
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR