Samarium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Samarium(III) i-propoxide
41311.AC
41311.AE
ALFA41311.ACEA
0
SGD
ALFA41311.AC
ALFA41311.AE
Samarium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99% (REO, rare earth oxide basis)
Samarium(III) isopropoxide
Purity: 99%
CAS No.: 3504-40-3
Molecular Formula: C9H21O3Sm
Formula Weight: 327.50
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Odour: Alcohol-like
UN No.: UN3274
MDL No.: MFCD00058820
Applications: Precursor for CVD and Sol-Gel
Formula:
C₉H₂₁O₃Sm MW: 327,621 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00058820 CAS Number: 3504-40-3 UN: 3274 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...