Bis(hexafluoroacetylacetonato)cobalt(II) hydrate ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Hexafluoroacetylacetono Cobalt(II) Salt, Cobalt(II) Hexafluoroacetylacetonate
41594.03
41594.06
ALFA41594.03EA
0
SGD
ALFA41594.03
ALFA41594.06
Bis(hexafluoroacetylacetonato)cobalt(II) hydrate ≥97%
Bis(hexafluoroacetylacetonato)cobalt(II) hydrate
Purity: 97%
CAS No.: 19648-83-0
Molecular Formula: C10H2CoF12O4.xH2O
Formula Weight: 473.04(anhy)
Physical Form: Powder
Appearance: Orange
Odour: Odourless
Melting Point: 173-179°
MDL No.: MFCD00042510
Formula:
C₁₀H₂CoF₁₂O₄ MW: 473,04 g/mol Melting Pt: 173…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042510 CAS Number: 19648-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...