Ammonium iron(III) oxalate hydrate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ferric ammonium oxalate
42112.18
42112.30
ALFA42112.18EA
0
SGD
ALFA42112.18
ALFA42112.30
Ammonium iron(III) oxalate hydrate
Ammonium iron(III) oxalate hydrate
CAS No.: 13268-42-3
Molecular Formula: (NH4)3Fe(C2O4)3.xH2O
Formula Weight: 374.02(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive; Hygroscopic
Melting Point: dec.
Solubility: Very soluble in water. Insoluble in alcohol
UN No.: UN3288
MDL No.: MFCD00013071
Applications: In photography, blueprints
Formula:
(NH₄)₃Fe(C₂O₄)₃·xH₂O MW: 374,02 g/mol (anhy) Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00013071 CAS Number: 13268-42-3 ADR: 6.1,III Merck Index: 14,517 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...