2-Naphthyl sulphate potassium salt ≥98%

Supplier: Thermo Fisher Scientific

44061.03 44061.MD
ALFA44061.03EA 0 SGD
ALFA44061.03 ALFA44061.MD
2-Naphthyl sulphate potassium salt ≥98%
2-Naphthyl sulphate potassium salt

Purity: 98% CAS No.: 1733-89-7 Molecular Formula: C10H7KO4S Formula Weight: 262.33 Physical Form: Powder Melting Point: 300° MDL No.: MFCD00004059

Formula: C₁₀H₇KO₄S
MW: 262.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004059
CAS Number: 1733-89-7
EINECS: 217-072-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR