(-)-O-Acetyl-L-malic anhydride ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-2-Acetoxysuccinic Anhydride, (S)-(-)-2-Acetoxysuccinic Anhydride
44366.09
44366.18
ALFA44366.09EA
0
SGD
ALFA44366.09
ALFA44366.18
(-)-O-Acetyl-L-malic anhydride ≥98%
(-)-O-Acetyl-L-malic anhydride
Purity: 98%
CAS No.: 59025-03-5
Molecular Formula: C6H6O5
Formula Weight: 158.11
Physical Form: Powder
Sensitivity: Moisture Sensitive
Melting Point: 56-58°
MDL No.: MFCD00082708
Formula:
C₆H₆O₅ MW: 158,11 g/mol Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082708 CAS Number: 59025-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...