Allyl acrylate ≥95% stabilised
Supplier: Thermo Scientific
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
44463.14
44463.22
ALFA44463.14EA
0
SGD
ALFA44463.14
ALFA44463.22
Allyl acrylate ≥95% stabilised
Allyl acrylate
Stab. with 4-methoxyphenol
Purity: 95%
CAS No.: 999-55-3
Molecular Formula: CH2=CHCOOCH2CH=CH2
Formula Weight: 112.13
Flash Point: 32°(89°F)
Physical Form: Liquid
Density: 0.935
Refractive Index: 1.4360
Odour: Pungent
Boiling Point: 46-47°/40mm
Beilstein Reference: 1743576
UN No.: UN2929
MDL No.: MFCD00014949
Formula:
C₆H₈O₂ MW: 112,13 g/mol Boiling Pt: 46…47 °C (40 mmHg) Density: 0,935 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014949 CAS Number: 999-55-3 EINECS: 213-662-2 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...