2-Bromo-3-(2-tetrahydropyranyloxy)pyridine ≥97%
Supplier: Thermo Scientific
|
Warning
|
44774.03
44774.06
ALFA44774.03EA
0
SGD
ALFA44774.03
ALFA44774.06
2-Bromo-3-(2-tetrahydropyranyloxy)pyridine ≥97%
2-Bromo-3-(2-tetrahydropyranyloxy)pyridine
Purity: 97%
CAS No.: 93560-60-2
Molecular Formula: C10H12BrNO2
Formula Weight: 258.12
Physical Form: Solid
Melting Point: 67°
MDL No.: MFCD06411700
Formula:
C₁₀H₁₂BrNO₂ MW: 258,12 g/mol Melting Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD06411700 CAS Number: 93560-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...