Lithium DL-2-aminoadipate ≥97%
Supplier: Thermo Scientific
|
Warning
|
44797.02
44797.04
ALFA44797.02EA
0
SGD
ALFA44797.02
ALFA44797.04
Lithium DL-2-aminoadipate ≥97%
Lithium DL-2-aminoadipate
Purity: 97+%
Molecular Formula: C6H9Li2NO4
Formula Weight: 173.02
Physical Form: Powder
Appearance: White
MDL No.: MFCD06657665
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...