Lithium-2,2-di-n-propylacetate ≥97%
Supplier: Thermo Scientific
|
Warning
|
44798.02
44798.04
44798.09
ALFA44798.02EA
0
SGD
ALFA44798.02
ALFA44798.04
ALFA44798.09
Lithium-2,2-di-n-propylacetate ≥97%
Lithium-2,2-di-n-propylacetate
Purity: 97+%
Molecular Formula: LiO2CCH(CH2CH2CH3)2
Formula Weight: 150.14
Physical Form: Powder
MDL No.: MFCD04113112
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...