Zeolite ZSM-5, ammonium

Supplier: Thermo Scientific

45882.14 45882.22 45882.36
ALFA45882.14EA 0 SGD
ALFA45882.14 ALFA45882.22 ALFA45882.36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 425 m2/g, 80:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support

Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR