Polyethylene glycol methyl ether methacrylate, M.W. 13,000
Supplier: Thermo Scientific
|
Danger
|
46513.MD
46513.03
46513.06
ALFA46513.MDEA
0
SGD
ALFA46513.MD
ALFA46513.03
ALFA46513.06
Polyethylene glycol methyl ether methacrylate, M.W. 13,000
Polyethylene glycol methyl ether methacrylate
CAS No.: 26915-72-0
Molecular Formula: CH3(OCH2CH2)nOCOC(=CH2)CH3
Storage Temperature: 2°C to 8°C
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00241432
Formula:
CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃ |
MDL Number:
MFCD00241432 CAS Number: 26915-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...