Polyethylene glycol monomethyl ether-p-toluenesulphonate, M.W. 5,000
Supplier: Thermo Scientific
|
Warning
|
46824.03
46824.06
ALFA46824.03EA
0
SGD
ALFA46824.03
ALFA46824.06
Polyethylene glycol monomethyl ether-p-toluenesulphonate, M.W. 5,000
Polyethylene glycol monomethyl ether-p-toluenesulphonate
CAS No.: 58320-73-3
Molecular Formula: CH3(OCH2CH2)nOSO2C6H4CH3
Physical Form: Powder
Appearance: White to off-white
Formula:
CH₃(OCH₂CH₂)nOSO₂C₆H₄CH₃ Storage Temperature: Ambient |
CAS Number:
58320-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...