Triisopropoxyvanadium(V) oxide ≥96%

Supplier: Thermo Scientific
Warning

Synonyms: Vanadium(V) Triisopropoxide Oxide, Vanadium (V) tri-i-propoxy oxide, Vanadium(V) Oxytriisopropoxide, VTIP

89798.03 89798.06 89798.14 89798.22
ALFA89798.03EA 56.7 SGD
ALFA89798.03 ALFA89798.06 ALFA89798.14 ALFA89798.22
Triisopropoxyvanadium(V) oxide ≥96%
Triisopropoxyvanadium(V) oxide

Purity: 96% CAS No.: 5588-84-1 Molecular Formula: C9H21O4V Formula Weight: 242.20 Flash Point: 45°(113°F) Physical Form: Liquid Density: 1.04 Sensitivity: Moisture Sensitive Appearance: Pale yellow-orange Melting Point: -12° Boiling Point: 81-83°/2mm UN No.: UN1993 MDL No.: MFCD00015017

Formula: C₉H₂₁O₄V
MW: 244,20 g/mol
Boiling Pt: 81…83 °C (2 mmHg)
Melting Pt: –12 °C
Density: 1,04
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015017
CAS Number: 5588-84-1
EINECS: 226-997-4
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR