3-Methyl-1H-pyrazol-5(4H)-one ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-methyl-4,5-dihydro-1H-pyrazol-5-one

A10253.09 A10253.18 A10253.30
ALFAA10253.09EA 75.5 SGD
ALFAA10253.09 ALFAA10253.18 ALFAA10253.30
3-Methyl-1H-pyrazol-5(4H)-one ≥98%
3-Methyl-1H-pyrazol-5(4H)-one

Purity: 98+% CAS No.: 108-26-9 Molecular Formula: C4H6N2O Formula Weight: 98.11 Melting Point: 220-224° MDL No.: MFCD00020699

Formula: C₄H₆N₂O
MW: 98,11 g/mol
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00020699
CAS Number: 108-26-9
EINECS: 203-565-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR