2-Thiopheneacrylic acid predominantly trans ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (E)-3-(Thiophen-2-yl)acrylic acid, 3-(2-Thienyl)acrylic Acid

A10342.09 A10342.18
ALFAA10342.09EA 0 SGD
ALFAA10342.09 ALFAA10342.18
2-Thiopheneacrylic acid predominantly trans ≥98%
2-Thiopheneacrylic acid

Purity: 98+% CAS No.: 15690-25-2 Molecular Formula: C7H6O2S Formula Weight: 154.19 Melting Point: 145-148° Beilstein Reference: 112053 MDL No.: MFCD00005455

Formula: C₇H₆O₂S
MW: 154,18 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00005455
CAS Number: 15690-25-2
EINECS: 214-402-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR